missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-2C Adrenergic R/ADRA2C Antibody (S330A-80), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-22397-0.025mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
alpha-2C Adrenergic R/ADRA2C Monoclonal specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Spécification
| alpha-2C Adrenergic R/ADRA2C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 50kDa or larger (possibly due to dimerization). Does not cross-react with other adrenergic receptors. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S330A-80 | |
| Unconjugated | |
| P18825 | |
| ADRA2C | |
| Synthetic peptide amino acids 442- 462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor | |
| 0.025 mg | |
| GPCR, Neuroscience | |
| 152 | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu