missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 2 Mouse anti-Human, Clone: ACTN2/3294, Novus Biologicals™
Mouse Monoclonal Antibody
249.00€
Spezifikation
| Antigène | Alpha Actinin 2 |
|---|---|
| Clone | ACTN2/3294 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Beschreibung
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Spezifikation
| Alpha Actinin 2 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
| A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| ACTN2/3294 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 88 | |
| IgG2b κ | |
| Protein A or G purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts