missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Marque: Novus Biologicals NBP1-30894
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
alpha-Smooth Muscle Actin Polyclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Canine, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Spécification
| alpha-Smooth Muscle Actin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| ACTA2 | |
| Alpha-Smooth Muscle Actin Antibody is made to a carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Alpha Smooth Muscle Actin. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and Mouse and Rat. Cross rectivity determined by Western blot Not yet tested in any other species. Expected cross reactivity species (based on sequence identity): Chicken (100%), Pig (100%), Rabbit (100%), Bovine (100%), Rhesus Monkey (100%). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen Assay-dependent dilution., Knockdown Validated | |
| AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
| Rabbit | |
| 42 kDa | |
| 100 μL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| 59 | |
| Human, Mouse, Rat, Canine, Zebrafish | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu