missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (3C11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-18258CL1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Dot Blot,Immunocytochemistry/Immunofluorescence
Spezifikation
| alpha-Synuclein | |
| Monoclonal | |
| CoraFluor 1 | |
| alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein | |
| Human alpha-Synuclein monomer | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot, Dot Blot, Immunocytochemistry/Immunofluorescence | |
| 3C11 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction