missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOTL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23351-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Regulates the translocation of phosphorylated SRC to peripheral cell-matrix adhesion sites. Required for proper architecture of actin filaments. Inhibits the Wnt/beta-catenin signaling pathway, probably by recruiting CTNNB1 to recycling endosomes and hence preventing its translocation to the nucleus. Participates in angiogenesis. May play a role in the polarity, proliferation and migration of endothelial cells. Selectively promotes FGF-induced MAPK activation through SRC.Spécification
| AMOTL2 | |
| Polyclonal | |
| Unconjugated | |
| AMOTL2 | |
| AMOTL2, angiomotin like 2, Angiomotin like protein 2, KIAA0989, LCCP, Leman coiled coil protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51421 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2J4 | |
| AMOTL2 | |
| AMOTL2 Fusion Protein Ag19953 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu