missing translation for 'onlineSavingsMsg'
Learn More

AMPD3 Antibody (AMPD3/901) - Azide and BSA Free, Novus Biologicals™

Code produit 18178437 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mg
Conditionnement:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18178437 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18178437 Fournisseur Novus Biologicals Code fournisseur NBP2476910.1MG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

AMPD3 Monoclonal specifically detects AMPD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Spécification

Antigène AMPD3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone AMPD3/901
Concentration 1.0 mg/mL
Conjugué Unconjugated
Dilution Flow Cytometry : 1 - 2 ug/million cells in 0.1ml, Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Immunocytochemistry : 1 - 2.0 ug/ml, CyTOF-ready
Formule PBS with No Preservative
Alias de gène adenosine monophosphate deaminase (isoform E), adenosine monophosphate deaminase 3, AMP aminohydrolase, AMP deaminase 3, AMP deaminase isoform E, EC 3.5.4.6, Erythrocyte AMP deaminase, erythrocyte type AMP deaminase, erythrocyte-specific AMP deaminase, myoadenylate deaminase
Symboles de gène(s) AMPD3
Espèces hôtes Mouse
Immunogène Recombinant full-length human AMDP3 protein
Méthode de purification Protein A or G purified
Quantité 0.1 mg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 272
Spécificité du test It recognizes a protein of ∼90kDa, which is identified as Adenosine Monophosphate Deaminase, isoform E (AMPD3). It has 767 amino acids and is assigned an EC 3.5.4.6. It is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. AMPD3 gene encodes the erythrocyte (E) isoforms, whereas other family members encode isoforms that predominate in muscle (M) and liver (L) cells. This MAb shows reactivity with cells of the erythroid lineage at all stages of maturation in the peripheral blood, bone marrow, and fetal liver. Non-erythroid lineages are negative by flow cytometry. This MAb is useful in the diagnosis of erythroleukemia, identification of bone marrow erythroid precursors, gating erythroid nucleated precursor cells from malignant cells in bone marrow specimens.
Espèces cibles Human
Contenu et stockage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG2b κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.