missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Amphiphysin Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA582702
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: PGFLYKVETL HDFEAANSDE LTLQRGDVVL VVPSDSEADQ DAGWLVGVKE SDWLQYRDLA TYKGLFPENF TRRL Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
Amphiphysins, members of the BAR (Bin-Amphiphysin-Rvsp) protein super family, play a key role in clathrin-mediated endocytosis of synaptic vesicles (SVs). All members of the BAR family share a highly conserved N-terminal BAR domain and a C-terminal Src homology (SH3) domain. Two isoforms of amphiphysin have been identified and act in concert as a heterodimer. Amphiphysins interact via its carboxy-terminal SH3 domain with dynamin, synaptojanin and clathrin. These complexes are implicated in synaptic vesicle recycling at nerve terminal. Amphiphysins were also identified as the dominant autoantigen in paraneoplastic Stiff-Man Syndrome.
Spécification
| Amphiphysin | |
| Polyclonal | |
| Unconjugated | |
| Amph | |
| Amph; AMPH1; amphiphysin; amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen); amphiphysin 1; amphiphysin I; HGNC:471 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49418 | |
| Amph | |
| Recombinant protein corresponding to Human AMPH. Recombinant protein control fragment (Product #RP-92625). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu