missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 4/ANGPTL4 Antibody (Kairos-1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-80039APC
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Monoclonal | |
| APC | |
| angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| Recombinant human Angiopoietin-like Protein 4/ANGPTL4. | |
| 0.1 mL | |
| Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| 51129 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Kairos-1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu