missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD39 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24027-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
ANKRD39 | |
Polyclonal | |
Unconjugated | |
ANKRD39 | |
ANKRD39, ankyrin repeat domain 39 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51239 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53RE8 | |
ANKRD39 | |
ANKRD39 Fusion Protein Ag21210 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |