missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A2 Mouse anti-Human, Clone: 1C1E12, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Spécification
| Antigène | Annexin A2 |
|---|---|
| Clone | 1C1E12 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spécification
| Annexin A2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Annexin 2, annexin A2, Annexin II, ANX2, ANX2L4, ANXA2, CAL1H, Calpactin 1 heavy chain, Calpactin I heavy chain, Chromobindin 8, LIP2, Lipocortin II, LPC2, LPC2D, P36, PAP IV, Protein I | |
| ANXA2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1C1E12 | |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| P07355 | |
| 302 | |
| Annexin A2 Fusion Protein Ag16654 | |
| Primary | |
| -20°C | |
| ANXA2 |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit