Learn More
Rho guanine nucleotide exchange factor (GEF) 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ARHGEF3.
Marque: Abnova H00050650-A01
Description
Rho-like GTPases are involved in a variety of cellular processes, and they are activated by binding GTP and inactivated by conversion of GTP to GDP by their intrinsic GTPase activity. Guanine nucleotide exchange factors (GEFs) accelerate the GTPase activity of Rho GTPases by catalyzing their release of bound GDP. This gene encodes a guanine nucleotide exchange factor, which specifically activates two members of the Rho GTPase family: RHOA and RHOB, both of which have a role in bone cell biology. It has been identified that genetic variation in this gene plays a role in the determination of bone mineral density (BMD), indicating the implication of this gene in postmenopausal osteoporosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: EPSNKRVKPLSRVTSLANLIPPVKATPLKRFSQTLQRSISFRSESRPDILAPRPWSRNAAPSSTKRRDSKLWSETFDVCVNQMLTSKEIKRQEAIFELSQGEEDLIEDLKSpécification
| Rho guanine nucleotide exchange factor (GEF) 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ARHGEF3. | |
| ARHGEF3 | |
| DKFZp434F2429/FLJ98126/GEF3/MGC118905/STA3/XPLN | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| NM_019555 | |
| ARHGEF3 | |
| ARHGEF3 (NP_062455, 33 a.a. ∼ 142 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 50650 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.