Learn More
CD164 molecule, sialomucin, Mouse, Clone: 4B4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CD164.
Marque: Abnova H00008763-M10.100ug
Description
Sialomucins are a heterogeneous group of secreted or membrane-associated mucins that appear to play 2 key but opposing roles in vivo: first as cytoprotective or antiadhesive agents, and second as adhesion receptors. CD164 is a type I integral transmembrane sialomucin that functions as an adhesion receptor (Watt et al., 1998 [PubMed 9680353]; Forde et al., 2007 [PubMed 17077324]).[supplied by OMIM
Sequence: MSRLSRSLLWAATCLGVLCVLSADKNTTQHPNVTTLAPISNVTSAPVTSLPLVTTPAPETCEGRNSCVSCFNVSVVNTTCFWIECKDESYCSHNSTVSDCQVGNTTDFCSVSTATSpécification
| CD164 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC011522 | |
| CD164 | |
| CD164 (AAH11522, 1 a.a. ∼ 115 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Adhesion | |
| 8763 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
| ELISA, Western Blot | |
| 4B4 | |
| Mouse monoclonal antibody raised against a partial recombinant CD164. | |
| CD164 | |
| MGC-24/MUC-24/endolyn | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.