Learn More
CD1d Mouse anti-Human, Brilliant Violet 510, Clone: CD1d42 (also known as 42.1), BD
Mouse Monoclonal Antibody
224.40€ - 520.20€
Spécification
| Antigène | CD1d |
|---|---|
| Clone | CD1d42 (also known as 42.1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugué | Brilliant Violet 510 |
Description
The CD1d42 monoclonal antibody recognizes CD1d. Cell surface CD1d is structurally homologous to Class I MHC molecules. It consists of a glycosylated type I transmembrane α chain (43-49 kDa) that is non-covalently associated with β2-microglobulin. CD1d is a member of the CD1 family of molecules, which belong to the immunoglobulin superfamily. Sequence homology data classifies the CD1 molecules into two groups. Group 1 includes CD1a, CD1b and CD1c molecules; group 2 includes CD1d molecules and their homologs in other species. CD1d is expressed on cortical thymocytes, B cells, dendritic cells, monocytes, and some nonlymphoid cells including intestinal epithelial cells, hepatocytes and keratinocytes. It is not expressed on resting mature T cells. Studies suggest that CD1d participates in lipid antigen presentation to CD1d-restricted NKT cells.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.
Spécification
| CD1d | |
| Flow Cytometry | |
| Brilliant Violet 510 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| R3; R3G1; HMC class I antigen-like glycoprotein CD1D |
| CD1d42 (also known as 42.1) | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human CD1d Recombinant Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.