Learn More
CD229 Mouse anti-Human, PE, Clone: HLy9.1.25, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 565238
Description
The HLY9.1.25 monoclonal antibody specifically binds to CD229 which is also known as, Signaling lymphocyte activation molecule family member 3 (SLAMF3), Lymphocyte antigen 9 (Ly9), or T-lymphocyte surface antigen Ly-9. CD229 is a 100-120 kDa, single-pass type I membrane glycoprotein that belongs to the SLAM family within the Ig superfamily. CD229 is expressed on thymocytes, T cells, B cells, dendritic cells, and NK cells. Through homophilic binding, CD229 may play a role in adhesive interactions between T and B cells. During T cell activation, CD229 localizes to the immunological synapse.
Spécification
| CD229 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human hLy9/CD229 Transfected Cell Line | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HLy9.1.25 | |
| SLAMF3; SLAM family member 3; LY9; Ly-9; hly9 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.