missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse anti-Human, Unlabeled, Clone: CD28.2, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 556620

Code nomenclature Nacres: NA.46

Code produit. 15808758

  • ONLINE EXCLUSIVE PRICE
    180.00€
    179.76€ / 1 pièce
    Économisez 0.24€
    0.13% de réduction

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

The CD28.2 monoclonal antibody specifically binds to CD28, a 44 kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.

This clone also cross-reacts with a subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, rhesus and cynomolgus macaques. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes; however, as described in the literature, the frequency of CD28.2-positive lymphocytes is reduced in the non-human primate, with the majority of cells being CD28 negative.

TRUSTED_SUSTAINABILITY
Spécification

Spécification

CD28
Monoclonal
0.5mg/mL
CD28 antigen; T44; Tp44; TP44
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Flow Cytometry
CD28.2
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Human CD28 Transfected Cell Line
0.1 mg
Immunology
Baboon, Cynomolgus, Rhesus Monkey
IgG1 κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis

For Research Use Only.