Learn More
CD29 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: HM beta1-1, BD
Hamster Monoclonal Antibody
Marque: BD Biosciences 562219
Description
The HM β1-1 monoclonal antibody specifically binds to the 130-kDa integrin β1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin-α chains. With α1 through α6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with αv (CD51), it forms αα8, and α9 chains in non-lymphoid tissues. As a result, CD29 has a broad tissue distribution, including lymphocytes, endothelia, smooth muscle, epithelia, and oocytes. This hamster mAb to a mouse leukocyte antigen has been observed to crossreact with similar populations of rat leukocytes. Source of the immunogen was purified mouse VLA-4 (α4β1, CD49d/CD29).
Spécification
| CD29 | |
| Monoclonal | |
| 1.0mg/mL | |
| Itgb1; Integrin beta-1; Fnrb; gpIIa; VLA-4 subunit beta | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 |
| Flow Cytometry | |
| HM beta1-1 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Purified mouse VLA-4 | |
| 0.5 mg | |
| Primary | |
| Store undiluted at 4°C |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.