Learn More
CD32 Mouse anti-Human, FITC, Clone: FLI8.26, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 555448
Description
The FLI8.26 monoclonal antibody specifically recognizes CD32 which is also known as Fcγreceptor type II (FcγRII). CD32 is a type I transmembrane glycoprotein that serves as a low affinity receptor for aggregated IgG. The FLI8.26 antibody recognizes a, b, and c forms of CD32 that are encoded by separate genes: CD32a/ FcγRIIa ( FCGR2A ), CD32b/ FcγRIIb ( FCGR2B ). These forms are differentially expressed on monocytes, granulocytes, T cells, B cells, NK cells, or platelets. These receptors play various roles in mediating and regulating inflammation and immunity including phagocytosis, cytotoxicity, degranulation, or platelet activation.
Spécification
| CD32 | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| K562 or FcγRII+ L cell lines | |
| 100 Tests | |
| Immunology | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| IgG2b κ |
| Flow Cytometry | |
| FLI8.26 | |
| CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.