missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD36 PE, Clone: 72-1, Invitrogen™
CD36 Monoclonal antibody A16204 from Invitrogen specifically detects CD36 in Mouse samples. It is validated for Flow Cytometry.
Marque: Invitrogen A16204
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD36 Monoclonal antibody A16204 from Invitrogen specifically detects CD36 in Mouse samples. It is validated for Flow Cytometry.
CD36 is a cell surface class B scavenger receptor. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signaling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. It may function as a cell adhesion molecule and directly mediates cyto- adherence of Plasmodium falciparum parasitized erythrocytes. CD36 binds long chain fatty acids and may function in the transport and/or as a regulator of fatty acid transport.Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2a, κ
Clone: 72-1
Immunogen: Mouse CD36 Protein
Formulation: 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV
Uniprot Accession: Q08857
Entrez GeneID: 12491
Gene Symbol: Cd36
This product detects CD36 in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse CD36 Protein.
Spécification
| CD36 | |
| Monoclonal | |
| PE | |
| Q08857 | |
| Cd36 | |
| Mouse CD36 Protein | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 72-1 | |
| 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide | |
| FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV | |
| Rat | |
| 50 μg | |
| 12491 | |
| Store at 4° C | |
| IgG2a κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu