Learn More
CDC14 cell division cycle 14 homolog A (S. cerevisiae), Mouse, Clone: 1F11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CDC14A.
Marque: Abnova H00008556-M02.100ug
Description
The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms. [provided by RefSeq
Sequence: PFRLSSSLQGSAVTLKTSKMALSPSATAKRINRTSLSSGATVRSFSINSRLASSLGNLNAATDDPENKKTSSSSKAGFTASPFTNLLNGSSQPTTRNYPESpécification
| CDC14 cell division cycle 14 homolog A (S. cerevisiae) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003672 | |
| CDC14A | |
| CDC14A (NP_003663, 431 a.a. ∼ 530 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1F11 | |
| Mouse monoclonal antibody raised against a partial recombinant CDC14A. | |
| CDC14A | |
| cdc14/hCDC14 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 8556 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.