missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hexamethylene bis-acetamide inducible 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HEXIM1 protein.
Marque: Abnova H00010614-B01.50uL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. This gene has no introns. [provided by RefSeq
Sequence: MAEPFLSEYQHQPQTSNCTGAAAVQEELNPERPPGAEERVPEEDSRWQSRAFPQLGGRPGPEGEGSLESQPPPLQTQACPESSCLREGEKGQNGDDSSAGGDFPPPAEVEPTPEAELLAQPCHDSEASKLGAPAAGGEEEWGQQQRQLGKKKHRRRPSKKKRHWKPYYKLTWEEKKKFDEKQSLRASRIRAEMFAKGQPVAPYNTTQFLMDDHDQEEPDLKTGLYSKRAAAKSDDTSDDDFMEEGGEEDGGSDGMGGDGSEFLQRDFSETYERYHTESLQNMSKQELIKEYLELEKCLSRMEDENNRLRLESKRLGGDDARVRELELELDRLRAENLQLLTENELHRQQERAPLSKFGDSpécification
| hexamethylene bis-acetamide inducible 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human HEXIM1 protein. | |
| HEXIM1 | |
| CLP1/EDG1/FLJ13562/HIS1/MAQ1 | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC006460 | |
| HEXIM1 | |
| HEXIM1 (AAH06460, 1 a.a. ∼ 359 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 10614 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu