Learn More
HLA-DR Mouse anti-Human, APC, Clone: L243 (G46-6), BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 559866
Description
Reacts with HLA-DR, a human class II antigen II of the major histocompatibility complex (MHC). HLA-DR is a transmembrane glycoprotein composed of an α chain (36kD) and a β subunit (27kD) expressed primarily on antigen presenting cells: B cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in cellular interaction during antigen presentation.
Spécification
| HLA-DR | |
| Monoclonal | |
| APC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
| IgG2a κ |
| Flow Cytometry | |
| L243 (G46-6) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.