Learn More
HLA-G Mouse anti-Human, Unlabeled, Clone: 4H84, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 557577
Description
HLA-G is a HLA class I molecule, but it is distinct from other class I molecules because of its quasi-monomorphism and restricted tissue distribution. It can be expressed as seven distinct protein forms. HLA-G1, -G2, G3, and -G4 are membrane bound while HLA-G5, -G6, and G7 are soluble. HLA-G is expressed in healthy individuals on amniocytes and cytotrophoblasts in the amnion-chorion. HLA-G interacts with inhibitory receptors such as ILT-2, ILT-4, p49 and KIR2DL4 expressed on NK cells, shifts the cytokine balance towards Th2 dominance, suppresses the proliferation of allogeneic CD4 lymphocytes and, in its soluble form, induces apoptosis of activated CD8 cells and inhibits cytolysis by NK cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 4H84
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Spécification
| HLA-G | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 4H84 | |
| Unconjugated | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.