Learn More
Hsp60 Mouse, Unlabeled, Clone: 24/HSP60, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 611563
Description
Heat shock proteins (Hsp) are a set of highly conserved proteins that include constitutively expressed (Hsp60, Hsp70, and Hsp90) and stress-induced (Hsp27 and Hsp72) proteins. Hsp60 is localized to the mitochondria, where it promotes mitochondrial protein folding and facilitates proteolytic degradation of misfolded or denatured proteins. It binds Hsp10, which regulates the substrate binding and ATPase activity of Hsp60. In HeLa and Jurkat mitochondria, Hsp60 associates with caspase-3 to form a complex that dissociates and releases from the mitochondria during apoptosis. In addition, Hsp60 accelerates the maturation of procaspase-3 through its ATP-dependent “foldase” activity. In addition to its role in protein folding, Hsp60 has also been implicated in immune function. In macrophages, its binding to the toll-like receptor-4 complex induces production of TNFα and nitric oxide and stimulation of a proinflammatory response. Thus, the protein folding function of Hsp60 is involved in mitochondrial protein folding in both normal and apoptotic cells, while release of Hsp60 during necrosis is thought to stimulate a proinflammatory response.
Host Species: Mouse
Clone: 24
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Hsp60 aa. 1-200
Formula Weight [Chemical]: 60kDa
Immunofluorescence, Western Blotting
Spécification
| Hsp60 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human Hsp60 aa. 1-200 | |
| 150 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 24/HSP60 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.