missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV Type 16 E2 Mouse anti-Virus, Clone: TVG 271, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA190530
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA1-90530 detects HPV16 E2 from virus samples. MA1-90530 has been successfully used in Western blot applications. The MA1-90530 immunogen is: Maltose binding protein MBP-E2 (generated from an expression vector in which the E2 ORF was cloned in frame with malE).
The human papilloma virus (HPV) family of DNA tumor viruses includes HPV-16 and HPV-18, which are associated with a large proportion of cervical cancer cases. E1 and E2 are proteins involved in the regulation of viral DNA replication. Antibodies to this target are useful for detection of protein in cervical lesions.
Spécification
| HPV Type 16 E2 | |
| Monoclonal | |
| 1.1 mg/mL | |
| HPV16; HPV-16; HPV E2 | |
| Maltose binding protein MBP-E2 (generated from an expression vector in which the E2 ORF was cloned in frame with malE). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| TVG 271 | |
| Unconjugated | |
| Mouse | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu