missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1, κ, Rat, Alexa Fluor™ 488, Clone: R3-34, BD
443.00€ - 452.00€
Spécification
Clone | R3-34 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugué | Alexa Fluor 488 |
Espèces hôtes | Rat |
Isotype | IgG1 κ |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15879008
|
BD Biosciences
557720 |
0.1 mg |
452.00€
1 pièce |
Expédition estimée: 04-06-2024
Connectez-vous pour voir le stock disponible |
|||||
15849078
|
BD Biosciences
557856 |
100 Tests |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1 κ
Flow Cytometry, Intracellular Staining, Isotype Control
Spécification
R3-34 | |
Alexa Fluor 488 | |
IgG1 κ | |
Undiluted at 4°C |
Flow Cytometry, Western Blot | |
Rat | |
Affinity Purified |