missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, κ, Rat, Alexa Fluor™ 488, Clone: R3-34, BD

Marque:  BD Biosciences 557856

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15849078

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
Voir les produits de remplacement »


Description

Description

The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: R3-34
Isotype: IgG1 κ

Flow Cytometry, Intracellular Staining, Isotype Control

Spécification

Spécification

Flow Cytometry, Western Blot
Alexa Fluor 488
Affinity Purified
Undiluted at 4°C
R3-34
Rat
100 Tests
IgG1 κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions