Learn More
leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human LILRB1 protein.
Marque: Abnova H00010859-D01P.100ug
Description
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MTPILTVLICLGLSLGPRTHVQAGHLPKPTLWAEPGSVITQGSPVTLRCQGGQETQEYRLYREKKTAPWITRIPQELVKKGQFPIPSITWEHAGRYRCYYGSDTAGRSESSDPLELVVTGAYIKPTLSAQPSPVVNSGGNVTLQCDSQVAFDGFILCKEGEDEHPQCLNSQPHARGSSRAIFSVGPVSPSRRWWYRCYAYDSNSPYEWSLPSDLLELLVLGVSKKPSLSVQPGPIVAPEETLTLQCGSDAGYNRFVLYKDGERDFLQLAGAQPQAGLSQANFTLGPVSRSYGGQYRCYGAHNLSSEWSAPSDPLDILIAGQFYDRVSLSVQPGPTVASGENVTLLCQSQGWMQTFLLTKEGAADDPWRLRSTYQSQKYQAEFPMGPVTSAHAGTYRCYGSQSSKPYLLTHPSDPLELVVSGPSGGPSSPTTGPTSTSGPEDQPLTPTGSDPQSGLGRHLGVVIGILVAVILLLLLLLLLFLILRHRRQGKHWTSTQRKADFQHPAGAVGPEPTDRGLQWRSSPAADAQEENLYAAVKHTQPEDGVEMDTRSPHDEDPQAVTYAEVKHSRPRREMASPPSPLSGEFLDTKDRQAEEDRQMDTEAAASEAPQDVTYAQLHSLTLRRKATEPPPSQEGPSPAVPSIYATLAIHSpécification
| leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human LILRB1 protein. | |
| LILRB1 | |
| CD85/CD85J/FLJ37515/ILT2/LIR-1/LIR1/MIR-7/MIR7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10859 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006669.2 | |
| LILRB1 | |
| LILRB1 (AAH15731.1, 1 a.a. ∼ 650 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.