missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKRN3 Monoclonal Antibody (2E10), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA520268
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Peptide Sequence: SFSAYWHQLV EPVRMGEGNM LYKSIKKELV VLRLASLLFK RFLSLRDELP FSEDQWDLLH YELEEYFNLI L
MKRN3 Monoclonal antibody specifically detects MKRN3 in Human samples. It is validated for ELISA, Immunocytochemistry.Spécification
MKRN3 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q13064 | |
MKRN3 | |
MKRN3 (NP_005655, 437 a.a. ∽ 507 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunocytochemistry | |
2E10 | |
Unconjugated | |
MKRN3 | |
CPPB2; D15S9; D7H15S9-1; makorin ring finger protein 3; makorin, ring finger protein, 3; Mkrn3; Probable E3 ubiquitin-protein ligase makorin-3; RING finger protein 63; RING-type E3 ubiquitin transferase makorin-3; RNF63; Zfp127; Zinc finger protein 127; ZNF127 | |
Mouse | |
Affinity chromatography | |
RUO | |
7681 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |