Learn More
nudix (nucleoside diphosphate linked moiety X)-type motif 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NUDT3 protein.
Marque: Abnova H00011165-B01.50uL
Description
NUDT3 belongs to the MutT, or Nudix, protein family. Nudix proteins act as homeostatic checkpoints at important stages in nucleoside phosphate metabolic pathways, guarding against elevated levels of potentially dangerous intermediates, like 8-oxo-dGTP, which promotes AT-to-CG transversions (Safrany et al., 1998 [PubMed 9822604]).[supplied by OMIM
Sequence: MMKLKSNQTRTYDGDGYKKRAACLCFRSESEEEVLLVSSSRHPDRWIVPGGGMEPEEEPSVAAVREVCEEAGVKGTLGRLVGIFENQERKHRTYVYVLIVTEVLEDWEDSVNIGRKREWFKIEDAIKVLQYHKPVQASYFETLRQGYSANNGTPVVATTYSVSAQSSMSGIRSpécification
| nudix (nucleoside diphosphate linked moiety X)-type motif 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human NUDT3 protein. | |
| NUDT3 | |
| DIPP/DIPP1 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_006703 | |
| NUDT3 | |
| NUDT3 (NP_006694, 1 a.a. ∼ 172 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 11165 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.