Learn More
2',5'-oligoadenylate synthetase 1, 40/46kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human OAS1 protein.
Marque: Abnova H00004938-D01P.100ug
Description
This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2'-specific nucleotidyl transfer reactions to synthesize 2',5'-oligoadenylates (2-5As). These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Mutations in this gene have been associated with host susceptibility to viral infection. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MMDLRNTPAKSLDKFIEDYLLPDTCFRMQINHAIDIICGFLKERCFRGSSYPVCVSKVVKGGSSGKGTTLRGRSDADLVVFLSPLTTFQDQLNRRGEFIQEIRRQLEACQRERAFSVKFEVQAPRWGNPRALSFVLSSLQLGEGVEFDVLPAFDALGQLTGGYKPNPQIYVKLIEECTDLQKEGEFSTCFTELQRDFLKQRPTKLKSLIRLVKHWYQNCKKKLGKLPPQYALELLTVYAWERGSMKTHFNTAQGFRTVLELVINYQQLCIYWTKYYDFKNPIIEKYLRRQLTKPRPVILDPADPTGNLGGGDPKGWRQLAQEAEAWLNYPCFKNWDGSPVSSWILLAESNSADDETDDPRRYQKYGYIGTHEYPHFSHRPSTLQAASTPQAEEDWTCTILSpécification
| 2',5'-oligoadenylate synthetase 1, 40/46kDa | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human OAS1 protein. | |
| OAS1 | |
| IFI-4/OIAS/OIASI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4938 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_016816.2 | |
| OAS1 | |
| OAS1 (NP_058132.2, 1 a.a. ∼ 400 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.