Learn More
K(lysine) acetyltransferase 2B, Mouse, Clone: 5E11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PCAF.
Marque: Abnova H00008850-M04.100ug
Description
CBP and p300 are large nuclear proteins that bind to many sequence-specific factors involved in cell growth and/or differentiation, including c-jun and the adenoviral oncoprotein E1A. The protein encoded by this gene associates with p300/CBP. It has in vitro and in vivo binding activity with CBP and p300, and competes with E1A for binding sites in p300/CBP. It has histone acetyl transferase activity with core histones and nucleosome core particles, indicating that this protein plays a direct role in transcriptional regulation. [provided by RefSeq
Sequence: LEEEVYSQNSPIWDQDFLSASSRTSQLGIQTVINPPPVAGTISYNSTSSSLEQPNAGSSSPACKASSGLEANPGEKRKMTDSHVLEEAKKPRVMGDIPMESpécification
| K(lysine) acetyltransferase 2B | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC060823 | |
| KAT2B | |
| PCAF (AAH60823, 353 a.a. ∼ 452 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 5E11 | |
| Mouse monoclonal antibody raised against a partial recombinant PCAF. | |
| KAT2B | |
| CAF/P/P/CAF/PCAF | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 8850 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.