Learn More
Invitrogen™ RSV strain RSS-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA14102
Description
PA1-4102 detects the respiratory syncytial virus strain RSS-2. PA1-4102 has been successfully used in Western blot procedures. The PA1-4102 immunogen is hRSV fusion protein of RSS-2 (subtype A) strain. This antibody reacts with residues K(201) Q L L P I V N K Q S(211) C of RSV F. Maintain at 4ºC for 3 months or at -20ºC for longer periods. Stable for 1 year. Avoid repeated freeze-thaw cycles.
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Spécification
| RSV strain RSS-2 | |
| Polyclonal | |
| Unconjugated | |
| human respiratory syncytial virus; respiratory syncytial virus strain rss-2; RSV; RSV RSS-2 | |
| hRSV fusion protein of RSS-2 (subtype A) strain. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.