Learn More
S100B Mouse, Unlabeled, Clone: 19, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 612376
Description
The S100 multigene family consists of a number of small dimeric molecules around 100 residues in length. The original members of this family, S100A and S100B, were initially identified in bovine brain. Other S100 proteins have also been identified, such as S100P from human placenta and S100L from bovine lung. The S100 family consists of at least 10 members which show a cell-type-specific expression pattern. Most S100 proteins are believed to be involved in Ca2+; signaling pathways. S100P consists of 95 amino acids and shares about 50% sequence identity with S100A and S100B, while S100L has been shown to possess 43-47% homology to S100A and S100B. S100B is expressed in the nervous system by glial cells, such as astrocyte, oligodendroglial, ependymal, and microglial cells. The dimeric form of S100B has been implicated in neuronal survival, neurite extension, and astrocyte growth. In addition, S100B expression is induced by stimulations that activate p53, and S100B activity may be involved in PKC-dependent p53 nuclear translocation in early G1 phase. Thus, S100B plays a role in calcium signaling pathways that regulate cell differentiation and cell cycle progression.
Immunofluorescence, Western Blotting
Spécification
| S100B | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse S100B aa. 1-92 | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
| In vitro Assay, Western Blot | |
| 19 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.