Learn More
sirtuin (silent mating type information regulation 2 homolog) 1 (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SIRT1.
Marque: Abnova H00023411-A01.50uL
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: NRYIFHGAEVYSDSEDDVLSSSSCGSNSDSGTCQSPSLEEPMEDESEIEEFYNGLEDEPDVPERAGGAGFGTDGDDQEAINEAISVKQEVTDMNYPSNKSSpécification
| sirtuin (silent mating type information regulation 2 homolog) 1 (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SIRT1. | |
| SIRT1 | |
| SIR2L1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC012499 | |
| SIRT1 | |
| SIRT1 (AAH12499, 456 a.a. ∼ 555 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 23411 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.