Learn More
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide, Mouse, Clone: 3G3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant YWHAG.
Brand: Abnova H00007532-M01.100ug
Beskrivelse
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways. [provided by RefSeq
Sequence: EQKTSADGNEKKIEMVRAYREKIEKELEAVCQDVLSLLDNYLIKNCSETQYESKVFYLKMKGDYYRYLAEVATGEKRATVVESSEKAYSEAHEISKEHMQTekniske data
| tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_012479 | |
| YWHAG | |
| YWHAG (NP_036611, 67 a.a. ∼ 166 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 7532 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a λ |
| ELISA | |
| 3G3 | |
| Mouse monoclonal antibody raised against a partial recombinant YWHAG. | |
| YWHAG | |
| 14-3-3GAMMA | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.