missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APE Antibody (13B8E5C2), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB100-116PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
APE Monoclonal antibody specifically detects APE in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Super Shift Assays, Proximity Ligation Assay, ChIP assay (ChIP), KnockDown
Spécification
| APE | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Purified |
| Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Supershift, Proximity Ligation Assay, ChIP Assay, KnockDown | |
| 13B8E5C2 | |
| Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP), Knockdown Validated | |
| AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease | |
| Purified human APE1 [Uniprot: P27695] | |
| 0.1 mL | |
| Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 328 | |
| Store at 4C in the dark. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu