missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5144460
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ARCA Polyclonal Antibody for Western Blot, ELISA
Member of the two-component regulatory system ArcB/ArcA. Represses a wide variety of aerobic enzymes under anaerobic conditions. Controls the resistance of E.coli to dyes; required for expression of the alkaline phosphatase and sex factor F genes; it may also be involved in the osmoregulation of envelope proteins. When activated by ArcB, it negatively regulates the expression of genes of aerobic function. Activates the transcription of the plfB operon by binding to its promoter.
Spécification
| ARCA | |
| Polyclonal | |
| Unconjugated | |
| arcA | |
| cpxC; dye; fexA; msp; seg; sfrA | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 69703137, 82262143 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.77 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P0A9Q1, P23793 | |
| arcA | |
| Recombinant Escherichia coli (strain K12) ARCA protein. | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu