Learn More
Arginase 1/ARG1/liver Arginase Antibody (ARG1/1125), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-47998H
Code nomenclature Nacres: NA.46
Code produit. 18637177
  • 605.00€ / 0.10mL
Description
Arginase 1/ARG1/liver Arginase Monoclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Arginase 1/ARG1/liver Arginase
Monoclonal
HRP
ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase
Recombinant human Arginase 1/ARG1/liver Arginase protein fragment (around aa11-97) (exact sequence is proprietary) (Uniprot: P05089)
0.1 mL
Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction
383
Store at 4°C in the dark.
IgG3 κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
ARG1/1125
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified