missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP11B Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5106949
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sequence homology: Cow: 85%; Dog: 77%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Rabbit: 83%; Rat: 85%.
ARHGAP11B is a Hominin-specific protein that promotes development and evolutionary expansion of the brain neocortex. Able to promote amplification of basal progenitors in the subventricular zone, producing more neurons during fetal corticogenesis. Does not possess GTPase activator activity.
Spécification
| ARHGAP11B | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP11B | |
| ARHGAP11B; B'-T; FAM7B1; family with sequence similarity 7, member B1; GAP (1-8); Protein FAM7B1; Rho GTPase activating protein 11B; rho GTPase-activating protein 11B; Rho-type GTPase-activating protein 11B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 89839 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q3KRB8 | |
| ARHGAP11B | |
| Synthetic peptide directed towards the C-terminal region of Human ARHGAP11B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu