Learn More
ARHI Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10811-2-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The ARHI antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARHI. This antibody recognizes human, mouse, rat antigen. The ARHI antibody has been validated for the following applications: IHC, ELISA analysis.Spécification
| ARHI | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9077 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, ELISA | |
| 600 μg/mL | |
| ARHI; NOEY2 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 26-30 kDa | |
| Liquid |