missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARL7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA556011
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: GIIYVVDSVD VDRLEEAKTE LHKVTKFAEN QGTPLLVIAN KQDLPKSLPV AEIEKQLALH ELIPATTYHV QPACAIIGEG LTEGMDKLYE MILKRRKSLK QKKKR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 64%.
ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport.
Spécification
| ARL7 | |
| Polyclonal | |
| Unconjugated | |
| ARL4C | |
| ADP ribosylation factor like GTPase 4C; ADP ribosylation factor-like protein 7; ADP-ribosylation factor like GTPase 4C; ADP-ribosylation factor-like 4C; ADP-ribosylation factor-like 7; ADP-ribosylation factor-like protein 4C; ADP-ribosylation factor-like protein 7; ADP-ribosylation factor-like protein LAK; ARL4C; ARL7; LAK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10123 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56559 | |
| ARL4C | |
| Recombinant protein corresponding to Human ARL7. Recombinant protein control fragment (Product #RP-88809). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu