missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13822-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognizes endogenous ARP3 and may cross-react with ATCR3B.
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.Spécification
| ARP3 | |
| Polyclonal | |
| Unconjugated | |
| ACTR3 | |
| Actin like protein 3, Actin related protein 3, ACTR3, ACTR3B, Arp11, ARP3, Arp4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10096, 74117, 81732 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61158, Q4V7C7, Q99JY9 | |
| Actr3 | |
| ARP3/ARP3B Fusion Protein Ag4922 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu