missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARSJ Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA557678
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: TADPYERVDL SNRYPGIVKK LLRRLSQFNK TAVPVRYPPK DPRSNPRLNG GVWGPWYKEE TKK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 52%.
ARSJ is a sulfatase that hydrolyzes sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. Sulfatases are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules.
Spécification
| ARSJ | |
| Polyclonal | |
| Unconjugated | |
| ARSJ | |
| 9330196J05Rik; Arsj; arylsulfatase family member J; arylsulfatase family, member J; arylsulfatase J; ASJ; D830047F08; RGD1307640; UNQ372/PRO708 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79642 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5FYB0 | |
| ARSJ | |
| Recombinant protein corresponding to Human ARSJ. Recombinant protein control fragment (Product #RP-104163). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu