missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA571961
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ASB3 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known.
Spécification
| ASB3 | |
| Polyclonal | |
| Unconjugated | |
| ASB3 | |
| 2400011J03Rik; ankyrin repeat and SOCS box containing 3; ankyrin repeat and SOCS box protein 3; ankyrin repeat and SOCS box-containing 3; ankyrin repeat and SOCS box-containing 3-like; ankyrin repeat and SOCS box-containing protein 3; ankyrin repeat-containing SOCS box protein 3; ASB3; Asb-3; EAR1; FLJ10123; FLJ10421; HREV; MGC12531; MGC132002; MGC996; THRAL | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 51130, 65257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9WV72, Q9Y575 | |
| ASB3 | |
| KLH conjugated synthetic peptide between 15-42 amino acids from the N-terminal region of human ASB3. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu