missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC/TMS1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-78978H
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative).
Spécification
| ASC/TMS1 | |
| Polyclonal | |
| Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
| Q9ULZ3 | |
| PYCARD | |
| This ASC/TMS1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [Uniprot: Q9ULZ3] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
| 29108 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| HRP | |
| PBS with No Preservative | |
| ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu