Learn More
Invitrogen™ ASC1 Recombinant Rabbit Monoclonal Antibody (PSH0-26)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA550797
Description
Positive control: A549 cell lysate, Hela cell lysate, U87-MG cell lysate, 293T cell lysate, HL-60 cell lysate, HepG2 cell lysate, RH-35 cell lysate, rat small intestine tissue. Predicted band size: 66 kDa Subcellular Location: Nucleus, Cytoplasm, cytosol, Cytoplasm, cytoskeleton, microtubule organizing center, centrosome.
Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.
Spécification
| ASC1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q15650 | |
| Trip4 | |
| Recombinant protein within human ASC1 aa 1-581/581. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| PSH0-26 | |
| Unconjugated | |
| Trip4 | |
| 4930558E03Rik; Activating signal cointegrator 1; Asc1; ASC-1; BB191711; HsT17391; SMABF1; thyroid hormone receptor interactor 4; thyroid receptor-interacting protein 4; transcription coactivator Asc1; TR-interacting protein 4; TRIP4; TRIP-4; ZC2HC5; zinc finger, C2HC5-type | |
| Rabbit | |
| Protein A | |
| RUO | |
| 315769, 9325 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.