Learn More
ASF/SF2 Mouse anti-Human, Mouse, Rat, Clone: 1A6C5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66671-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody can recognize all the 3 isoforms (28 kDa,33 kDa and 22 kDa) of SFRS1.
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.Spécification
| ASF/SF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q07955, Q6PDM2 | |
| SRSF1 | |
| ASF/SF2 Fusion Protein Ag3626 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1A6C5 | |
| Unconjugated | |
| SRSF1 | |
| Alternative splicing factor 1, ASF, ASF 1, ASF/SF2, SF2, SF2p33, SFRS1, SRp30a, SRSF1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 110809, 6426, 689890 | |
| -20°C | |
| Liquid |