Learn More
ASS1 Monoclonal Antibody (OTI4G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marque: Origene Technologies TA809138
Description
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.Spécification
| ASS1 | |
| Monoclonal | |
| Unconjugated | |
| ASS1 | |
| ASS, CTLN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 445 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4G9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00966 | |
| ASS1 | |
| Synthetic peptide corresponding to residues near C-terminus of human ASS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.