missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26723-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mitochondrial ATP synthase is composed of two multi-subunit complexes that utilize an inner membrane electrochemical gradient to catalyze the synthesis of ATP during oxidative phosphorylation. The two multi-subunit complexes are designated F1 and F0, the former of which comprises the soluble catalytic core and the latter of which comprises the membrane-spanning proton channel of ATP synthase. ATP8, also known as MT-ATP8, ATPASE8 or MTATP8, is a 68 amino acid single-pass membrane protein that localizes to mitochondria and functions as a minor subunit of the F0 complex. The gene encoding ATP8 maps to the mitochondrial chromosome and may be involved in the pathogenesis of multiple sclerosis.Spécification
ATP8 | |
Polyclonal | |
Unconjugated | |
ATP8 | |
A6L, ATP synthase protein 8, ATP8, ATPASE8, F ATPase subunit 8, MT ATP8, MTATP8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4509 | |
-20°C | |
Liquid |
Western Blot | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P03928 | |
ATP8 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |