missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE-1 Antibody (Genentech anti-BACE1), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals NBP3-28514CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| BACE-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
| BACE1 | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 23621 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech anti-BACE1 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu